Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylbenzimidazole, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1632-83-3
C8H8N2
132.17
MFCD00192275
FGYADSCZTQOAFK-UHFFFAOYSA-N
1-methyl-1h-benzimidazole,1h-benzimidazole, 1-methyl,methylbenzimidazole,unii-12lw89n19y,1-methyl-1h-benzo d imidazole,1h-benzimidazole, methyl,1-methyl-1,3-benzodiazole,1-methyl-1h-1,3-benzodiazole,pubchem7613,1-methylbenzimidazole
95890
1-methylbenzimidazole
CN1C=NC2=CC=CC=C21
1632-83-3
C8H8N2
132.17
MFCD00192275
FGYADSCZTQOAFK-UHFFFAOYSA-N
1-methyl-1h-benzimidazole,1h-benzimidazole, 1-methyl,methylbenzimidazole,unii-12lw89n19y,1-methyl-1h-benzo d imidazole,1h-benzimidazole, methyl,1-methyl-1,3-benzodiazole,1-methyl-1h-1,3-benzodiazole,pubchem7613,1-methylbenzimidazole
95890
1-methylbenzimidazole
CN1C=NC2=CC=CC=C21
Melting Point 55°C to 57°C
Boiling Point 154°C (16.0 hPa)
Color Pink to White
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 132.17
Physical Form Solid
Percent Purity 99%
Chemical Name or Material 1-Methylbenzimidazole
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning