Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylcyclohexanol, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
590-67-0
C7H14O
114.19
MFCD00003857
VTBOTOBFGSVRMA-UHFFFAOYSA-N
1-methylcyclohexanol,cyclohexanol, 1-methyl,1-methyl-1-cyclohexanol,cyclohexanol,methyl,1-methyl-cyclohexanol,1-methylcyclohexyl alcohol,acmc-1b1ai,1-methyl-cyclohexan-1-ol,1-methyl cyclohexanol
11550
1-methylcyclohexan-1-ol
CC1(CCCCC1)O
590-67-0
C7H14O
114.19
MFCD00003857
VTBOTOBFGSVRMA-UHFFFAOYSA-N
1-methylcyclohexanol,cyclohexanol, 1-methyl,1-methyl-1-cyclohexanol,cyclohexanol,methyl,1-methyl-cyclohexanol,1-methylcyclohexyl alcohol,acmc-1b1ai,1-methyl-cyclohexan-1-ol,1-methyl cyclohexanol
11550
1-methylcyclohexan-1-ol
CC1(CCCCC1)O
Specific Gravity 0.919
Flash Point 67°C
Melting Point 26°C
Boiling Point 168°C (752.0mmHg)
Refractive Index 1.4590 to 1.461
Infrared Spectrum Authentic
Density 0.9190g/mL
Quantity 5g
Packaging Glass bottle
Color Colorless to Yellow
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula CH3C6H10OH
Beilstein 06, 11
Formula Weight 114.19
Physical Form Liquid After Melting
Percent Purity 96%
Chemical Name or Material 1-Methylcyclohexanol
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Flammable solid.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/va

GHS Signal Word: Warning

EINECSNumber : 209-688-9