Alfa Aesar™ 1-Methylcyclopentanol, 98%

Manufacturer: Alfa Aesar™ B2474414

Quantity 25g

 View more versions of this product

Catalog No. AAB2474414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1462-03-9
    Molecular Formula C6H12O
    Formula Weight 100.16
    MDL Number MFCD00001364
    UN Number UN1325
    Chemical Name or Material 1-Methylcyclopentanol

    Specifications

    Formula Weight 100.16
    Molecular Formula C6H12O
    MDL Number MFCD00001364
    CAS 1462-03-9
    UN Number UN1325
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    Flash Point 40°C (105°F)
    Boiling Point 135° to 136°C
    Melting Point 36° to 37°C
    Quantity 25g
    Chemical Name or Material 1-Methylcyclopentanol
    Assay 98%

    GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit