Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylcyclopropanecarboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
6914-76-7
C5H8O2
100.12
MFCD00001290
DIZKLZKLNKQFGB-UHFFFAOYSA-N
1-methylcyclopropanecarboxylic acid,1-methyl-cyclopropanecarboxylic acid,cyclopropanecarboxylic acid, 1-methyl,cyclopropanecarboxylicacid, 1-methyl,pubchem16037,1-methylcyclopropanoic acid,acmc-209o6g,d07fpq,1-carboxy-1-methylcyclopropane,dizklzklnkqfgb-uhfffaoysa
81326
1-methylcyclopropane-1-carboxylic acid
CC1(CC1)C(=O)O
6914-76-7
C5H8O2
100.12
MFCD00001290
DIZKLZKLNKQFGB-UHFFFAOYSA-N
1-methylcyclopropanecarboxylic acid,1-methyl-cyclopropanecarboxylic acid,cyclopropanecarboxylic acid, 1-methyl,cyclopropanecarboxylicacid, 1-methyl,pubchem16037,1-methylcyclopropanoic acid,acmc-209o6g,d07fpq,1-carboxy-1-methylcyclopropane,dizklzklnkqfgb-uhfffaoysa
81326
1-methylcyclopropane-1-carboxylic acid
CC1(CC1)C(=O)O
Melting Point 34.0°C to 36.0°C
Quantity 5g
Packaging Glass bottle
Flash Point 84°C
Boiling Point 183.0°C to 185.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3C3H4CO2H
Beilstein 09, II, 5
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material 1-Methylcyclopropanecarboxylic acid
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 230-020-7