Alfa Aesar™ 1-Methylhydantoin, 97%

Manufacturer: Alfa Aesar™ B2432214

Quantity 25g

 View more versions of this product

Catalog No. AAB2432214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 616-04-6
    Molecular Formula C4H6N2O2
    Formula Weight 114.1
    MDL Number MFCD00003187
    Chemical Name or Material 1-Methylhydantoin

    Specifications

    Formula Weight 114.1
    Molecular Formula C4H6N2O2
    MDL Number MFCD00003187
    CAS 616-04-6
    Melting Point 156° to 159°C
    Quantity 25g
    Chemical Name or Material 1-Methylhydantoin
    Assay 97%

    Description

    Applications
    1-Methylhydantoin is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. 1-Methylhydantoin and 5-hydroxy-1-methylhydantoin were isolated as plant growth regulators from rabbit skin tissue which had been inflamed by inoculation with vaccinia virus.

    Solubility
    Soluble in methanol. (almost transparency). Insoluble in water.

    Notes
    Store at 4°C. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit