Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylhydantoin 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
616-04-6
C4H6N2O2
114.104
MFCD00003187
RHYBFKMFHLPQPH-UHFFFAOYSA-N
1-methylhydantoin,dioxy-creatinine,n-methylhydantoin,hydantoin, 1-methyl,1-methyl-hydantoin,methylhydantoin,n-methylimidazolidine-2,4-dione,unii-0546h9ur1r,1-methyldiazolidine-2,4-dione,2,4-imidazolidinedione, 1-methyl
69217
CHEBI:16354
1-methylimidazolidine-2,4-dione
CN1CC(=O)NC1=O
616-04-6
C4H6N2O2
114.104
MFCD00003187
RHYBFKMFHLPQPH-UHFFFAOYSA-N
1-methylhydantoin,dioxy-creatinine,n-methylhydantoin,hydantoin, 1-methyl,1-methyl-hydantoin,methylhydantoin,n-methylimidazolidine-2,4-dione,unii-0546h9ur1r,1-methyldiazolidine-2,4-dione,2,4-imidazolidinedione, 1-methyl
69217
CHEBI:16354
1-methylimidazolidine-2,4-dione
CN1CC(=O)NC1=O
Color White
Quantity 5g
Infrared Spectrum Authentic
Melting Point 156°C to 160°C
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 24, 244
Formula Weight 114.1
Physical Form Fine Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 1-Methylhydantoin, 97%
Show More Show Less

EINECSNumber : 210-460-6

TSCA : TSCA