Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1-Methylimidazole-4,5-dicarbonitrile, 97%
Click to view available options
Quantity:
1g
5g
25g
Specifications
Melting Point | 83°C to 86°C |
Quantity | 1g |
UN Number | UN3439 |
Formula Weight | 132.14 |
Percent Purity | 97% |
Chemical Name or Material | 1-Methylimidazole-4,5-dicarbonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures