Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methylimidazole-4,5-dicarbonitrile, 97%

Click to view available options
Quantity:
1g
5g
25g
19485-35-9
C6H4N4
132.126
MFCD01567299
SUOVGHDPXJUFME-UHFFFAOYSA-N
1-methyl-1h-imidazole-4,5-dicarbonitrile,4,5-dicyano-1-methyl-1h-imidazole,4,5-dicyano-1-methylimidazole,imidazole, 4,5-dicyano-1-methyl,1h-imidazole-4,5-dicarbonitrile, 1-methyl,pubchem8999,acmc-1bvri,1-methyl-4,5-dicyano-1h-imidazole,1h-imidazole-4,5-dicarbonitrile,1-methyl,1-methyl-1h-imidazole-4,5-dicarbonitrile #
583268
1-methylimidazole-4,5-dicarbonitrile
CN1C=NC(=C1C#N)C#N
19485-35-9
C6H4N4
132.126
MFCD01567299
SUOVGHDPXJUFME-UHFFFAOYSA-N
1-methyl-1h-imidazole-4,5-dicarbonitrile,4,5-dicyano-1-methyl-1h-imidazole,4,5-dicyano-1-methylimidazole,imidazole, 4,5-dicyano-1-methyl,1h-imidazole-4,5-dicarbonitrile, 1-methyl,pubchem8999,acmc-1bvri,1-methyl-4,5-dicyano-1h-imidazole,1h-imidazole-4,5-dicarbonitrile,1-methyl,1-methyl-1h-imidazole-4,5-dicarbonitrile #
583268
1-methylimidazole-4,5-dicarbonitrile
CN1C=NC(=C1C#N)C#N
Melting Point 83°C to 86°C
Quantity 5g
UN Number UN3439
Formula Weight 132.14
Percent Purity 97%
Chemical Name or Material 1-Methylimidazole-4,5-dicarbonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.