Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylindole-3-carboxaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
19012-03-4
C10H9NO
159.19
MFCD00014570
KXYBYRKRRGSZCX-UHFFFAOYSA-N
1-methyl-1h-indole-3-carbaldehyde,1-methylindole-3-carboxaldehyde,1-methyl-indole-3-carbaldehyde,1h-indole-3-carboxaldehyde, 1-methyl,3-formyl-1-methylindole,1-methyl-3-formylindole,n-methyl-3-formylindole,indole-3-carboxaldehyde, 1-methyl,3-formyl-1-methyl-1h-indole,n-methyl-3-indolecarboxaldehyde
87894
1-methylindole-3-carbaldehyde
CN1C=C(C2=CC=CC=C21)C=O
19012-03-4
C10H9NO
159.19
MFCD00014570
KXYBYRKRRGSZCX-UHFFFAOYSA-N
1-methyl-1h-indole-3-carbaldehyde,1-methylindole-3-carboxaldehyde,1-methyl-indole-3-carbaldehyde,1h-indole-3-carboxaldehyde, 1-methyl,3-formyl-1-methylindole,1-methyl-3-formylindole,n-methyl-3-formylindole,indole-3-carboxaldehyde, 1-methyl,3-formyl-1-methyl-1h-indole,n-methyl-3-indolecarboxaldehyde
87894
1-methylindole-3-carbaldehyde
CN1C=C(C2=CC=CC=C21)C=O
Melting Point 67°C to 72°C
Quantity 50g
Infrared Spectrum Authentic
Color Brown or Orange to Yellow
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 159.19
Physical Form Crystalline Powder and Chunks
Percent Purity ≥96.0%
Chemical Name or Material 1-Methylindole-3-carboxaldehyde
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.