Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methylindole-5-carboxaldehyde, 97%

Click to view available options
Quantity:
250mg
1g
90923-75-4
C10H9NO
159.188
MFCD08271908
XIVDZBIBWGQOTI-UHFFFAOYSA-N
1-methyl-1h-indole-5-carbaldehyde,1h-indole-5-carboxaldehyde, 1-methyl,5-formyl-1-methylindole,n-methylindol-5-carbaldehyde,n-methylindole-5-carbaldehyde,n-methylindole-5-carboxaldehyde,1-methyl-1h-indole-5-carboxaldehyde,1h-indole-5-carboxaldehyde,1-methyl,1-methyl-1h-indole-5-carboxaldehyde
7537534
1-methylindole-5-carbaldehyde
CN1C=CC2=C1C=CC(=C2)C=O
90923-75-4
C10H9NO
159.188
MFCD08271908
XIVDZBIBWGQOTI-UHFFFAOYSA-N
1-methyl-1h-indole-5-carbaldehyde,1h-indole-5-carboxaldehyde, 1-methyl,5-formyl-1-methylindole,n-methylindol-5-carbaldehyde,n-methylindole-5-carbaldehyde,n-methylindole-5-carboxaldehyde,1-methyl-1h-indole-5-carboxaldehyde,1h-indole-5-carboxaldehyde,1-methyl,1-methyl-1h-indole-5-carboxaldehyde
7537534
1-methylindole-5-carbaldehyde
CN1C=CC2=C1C=CC(=C2)C=O
Quantity 1g
Sensitivity Air & Moisture Sensitive
Beilstein 1448734
Formula Weight 159.18
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1-Methylindole-5-carboxaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning