Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylnaphthalene, 96%, Acros Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
90-12-0
C11H10
142.2
MFCD00004034
QPUYECUOLPXSFR-UHFFFAOYSA-N
alpha-methylnaphthalene,naphthalene, 1-methyl,methylnaphthalene,naphthalene, methyl,polymethylnaphthalene,1-methyl-naphthalene,alpha-methyl naphthalenes,methyl naphthalene,1-methyl naphthalene,fema number 3193
7002
CHEBI:50717
1-methylnaphthalene
CC1=CC=CC2=CC=CC=C12
90-12-0
C11H10
142.2
MFCD00004034
QPUYECUOLPXSFR-UHFFFAOYSA-N
alpha-methylnaphthalene,naphthalene, 1-methyl,methylnaphthalene,naphthalene, methyl,polymethylnaphthalene,1-methyl-naphthalene,alpha-methyl naphthalenes,methyl naphthalene,1-methyl naphthalene,fema number 3193
7002
CHEBI:50717
1-methylnaphthalene
CC1=CC=CC2=CC=CC=C12
Refractive Index 1.6130 to 1.6160
Flash Point 82°C
Color Colorless to Yellow
Specific Gravity 1.02
Infrared Spectrum Authentic
Melting Point -22.0°C
Packaging Glass bottle
Boiling Point 240.0°C to 243.0°C
Quantity 500g
Density 1.0200g/mL
CAS Min % 97+%
Assay Percent Range 96%
Linear Formula C10H7CH3
Beilstein 05, 566
Solubility Information Solubility in water: 26mg/l (25°C). Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 142.2
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 1-Methylnaphthalene, 97%
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 201-966-8

RTECSNumber : QJ9630000

TSCA : TSCA