Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpiperazine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
109-01-3
C5H12N2
100.16
PVOAHINGSUIXLS-UHFFFAOYSA-N
n-methylpiperazine,piperazine, 1-methyl,4-methylpiperazine,1-methyl-piperazine,n-methyl piperazine,1-methyl piperazine,methylpiperazine,n-methyl-piperazine,n-methylpiperazin,1-methylpyperazine
53167
1-methylpiperazine
CN1CCNCC1
109-01-3
C5H12N2
100.16
PVOAHINGSUIXLS-UHFFFAOYSA-N
n-methylpiperazine,piperazine, 1-methyl,4-methylpiperazine,1-methyl-piperazine,n-methyl piperazine,1-methyl piperazine,methylpiperazine,n-methyl-piperazine,n-methylpiperazin,1-methylpyperazine
53167
1-methylpiperazine
CN1CCNCC1
Viscosity 4 mPa.s (20°C)
Water 1% Max. (K.F.)
Specific Gravity 0.903
Packaging Glass bottle
Infrared Spectrum Authentic
Density 0.9030g/mL
Boiling Point 138.0°C
Color Undesignated
Quantity 2.5kg
Refractive Index 1.4645 to 1.4665
Melting Point -6.0°C
Flash Point 42°C
Assay Percent Range 98.5% min. (GC)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and aceton
Formula Weight 100.16
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1-Methylpiperazine
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 203-639-5