Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methylpiperidine-3-methanol, 98%

Click to view available options
Quantity:
50g
250g
7583-53-1
C7H15NO
129.203
MFCD00006497
UGXQXVDTGJCQHR-UHFFFAOYSA-N
1-methyl-3-piperidinemethanol,1-methylpiperidin-3-yl methanol,1-methylpiperidine-3-methanol,3-piperidinemethanol, 1-methyl,3-hydroxymethyl-1-methylpiperidine,1-methyl-3-piperidyl methanol,1-methyl-3-hydroxymethylpiperidine,1-methyl-piperidin-3-yl-methanol,3-piperidinemethanol,1-methyl-,--9ci
97998
(1-methylpiperidin-3-yl)methanol
CN1CCCC(C1)CO
7583-53-1
C7H15NO
129.203
MFCD00006497
UGXQXVDTGJCQHR-UHFFFAOYSA-N
1-methyl-3-piperidinemethanol,1-methylpiperidin-3-yl methanol,1-methylpiperidine-3-methanol,3-piperidinemethanol, 1-methyl,3-hydroxymethyl-1-methylpiperidine,1-methyl-3-piperidyl methanol,1-methyl-3-hydroxymethylpiperidine,1-methyl-piperidin-3-yl-methanol,3-piperidinemethanol,1-methyl-,--9ci
97998
(1-methylpiperidin-3-yl)methanol
CN1CCCC(C1)CO
Flash Point 94°C (201°F)
Quantity 50g
Boiling Point 140°C to 145°C
Beilstein 103713
Formula Weight 129.2
Percent Purity 98%
Chemical Name or Material 1-Methylpiperidine-3-methanol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning