Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpiperidine-4-carbothioamide, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
88654-17-5
C7H14N2S
158.263
MHYXWOIIIZRCAJ-UHFFFAOYSA-N
4-piperidinecarbothioamide, 1-methyl,acmc-20c53p,methylpiperidine-4 carbothioamide
15584501
1-methylpiperidine-4-carbothioamide
CN1CCC(CC1)C(=S)N
88654-17-5
C7H14N2S
158.263
MHYXWOIIIZRCAJ-UHFFFAOYSA-N
4-piperidinecarbothioamide, 1-methyl,acmc-20c53p,methylpiperidine-4 carbothioamide
15584501
1-methylpiperidine-4-carbothioamide
CN1CCC(CC1)C(=S)N
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color Tan to White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 158.27
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 1-Methylpiperidine-4-carbothioamide
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger