Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpyrazole, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
930-36-9
C4H6N2
82.1
MFCD00144943
UQFQONCQIQEYPJ-UHFFFAOYSA-N
1-methyl-1h-pyrazole,n-methylpyrazole,1h-pyrazole, 1-methyl,pyrazole, 1-methyl,methylpyrazole,1-methyl pyrazole,1-methypyrazole,n-methyl pyrazole,1-methyl-pyrazole,zlchem 255
70255
CHEBI:59025
1-methylpyrazole
CN1C=CC=N1
930-36-9
C4H6N2
82.1
MFCD00144943
UQFQONCQIQEYPJ-UHFFFAOYSA-N
1-methyl-1h-pyrazole,n-methylpyrazole,1h-pyrazole, 1-methyl,pyrazole, 1-methyl,methylpyrazole,1-methyl pyrazole,1-methypyrazole,n-methyl pyrazole,1-methyl-pyrazole,zlchem 255
70255
CHEBI:59025
1-methylpyrazole
CN1C=CC=N1
Boiling Point 127°C
Flash Point 35°C
Specific Gravity 0.993
Density 0.9930g/mL
Refractive Index 1.4760 to 1.478
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 50g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 82.1
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Methylpyrazole
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning