Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Methylpyrrolidine, 98%, Alfa Aesar™
Click to view available options
Quantity:
100g
500g
Description
Applications
1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.
Solubility
Miscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.
Solubility
Miscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications
Flash Point | -18°C (0°F) |
Refractive Index | 1.4247 |
Melting Point | °C to 90°C |
Boiling Point | 80°C to 81°C |
Density | 0.819 |
Quantity | 100g |
Assay Percent Range | 0.98 |
UN Number | UN3286 |
Solubility Information | Miscible with water. |
Formula Weight | 85.15 |
Show More |
Safety and Handling
GHS H Statement
H225-H314-H318-H302-H332
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H301-H314-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
EINECSNumber : 204-438-5
RTECSNumber : UY1420500
TSCA : Yes
Recommended Storage : Ambient temperatures