Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpyrrolidine, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic drum
120-94-5
C5H11N
85.15
AVFZOVWCLRSYKC-UHFFFAOYSA-N
n-methylpyrrolidine,methylpyrrolidine,n-methyltetrahydropyrrole,pyrrolidine, 1-methyl,n-methyl pyrrolidine,1-methyl-pyrrolidine,pyrrolidine, n-methyl,unii-06509tzu6c,methyl pyrrolidine
8454
1-methylpyrrolidine
CN1CCCC1
120-94-5
C5H11N
85.15
AVFZOVWCLRSYKC-UHFFFAOYSA-N
n-methylpyrrolidine,methylpyrrolidine,n-methyltetrahydropyrrole,pyrrolidine, 1-methyl,n-methyl pyrrolidine,1-methyl-pyrrolidine,pyrrolidine, n-methyl,unii-06509tzu6c,methyl pyrrolidine
8454
1-methylpyrrolidine
CN1CCCC1
Specific Gravity 0.819
Viscosity 0.48 mPa.s (20°C)
Flash Point −21°C
Density 0.8190g/mL
Boiling Point 80.0°C to 81.0°C
Quantity 500g
Melting Point -90.0°C
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4237 to 1.4257
Assay Percent Range 98%
Beilstein 20,4
Solubility Information Solubility in water: fully miscible.
Formula Weight 85.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Methylpyrrolidine
Show More Show Less