Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methylthio-1-methylamino-2-nitroethylene, 97%

Click to view available options
Quantity:
10g
50g
250g
61832-41-5
C4H8N2O2S
148.18
MFCD00040994
YQFHPXZGXNYYLD-ONEGZZNKSA-N
n-methyl-1-methylthio-2-nitroethenamine,unii-a9ls0uv4e3,ethenamine, n-methyl-1-methylthio-2-nitro,a9ls0uv4e3,n-methyl-1-methylthio-2-nitro-1-etheneamine,1-methylamino-1-methylthio-2-nitroethylene,e-n-methyl-1-methylthio-2-nitroethenamine,e-n-methyl-1-methylsulfanyl-2-nitroethenamine
3035401
(E)-N-methyl-1-methylsulfanyl-2-nitroethenamine
CNC(=C[N+](=O)[O-])SC
61832-41-5
C4H8N2O2S
148.18
MFCD00040994
YQFHPXZGXNYYLD-ONEGZZNKSA-N
n-methyl-1-methylthio-2-nitroethenamine,unii-a9ls0uv4e3,ethenamine, n-methyl-1-methylthio-2-nitro,a9ls0uv4e3,n-methyl-1-methylthio-2-nitro-1-etheneamine,1-methylamino-1-methylthio-2-nitroethylene,e-n-methyl-1-methylthio-2-nitroethenamine,e-n-methyl-1-methylsulfanyl-2-nitroethenamine
3035401
(E)-N-methyl-1-methylsulfanyl-2-nitroethenamine
CNC(=C[N+](=O)[O-])SC
Melting Point 109°C to 113°C
Quantity 50g
Beilstein 2828017
Formula Weight 148.18
Percent Purity 97%
Chemical Name or Material 1-Methylthio-1-methylamino-2-nitroethylene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning