Alfa Aesar™ 1-Methylthio-2-propanone, 98+%

Manufacturer: Alfa Aesar™ B2414922

Quantity 100g

 View more versions of this product

Catalog No. AAB2414922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14109-72-9
    Molecular Formula C4H8OS
    Formula Weight 104.17
    MDL Number MFCD00015325
    UN Number UN1224
    Beilstein 1737738
    Synonym Acetonyl methyl sulfide; (Methylthio)acetone
    Chemical Name or Material 1-Methylthio-2-propanone

    Specifications

    Formula Weight 104.17
    Molecular Formula C4H8OS
    MDL Number MFCD00015325
    CAS 14109-72-9
    UN Number UN1224
    Synonym Acetonyl methyl sulfide; (Methylthio)acetone
    Health Hazard 1 Warning
    Boiling Point 69° to 70°C (30mmHg)
    Quantity 100g
    Flash Point 43°C (109°F)
    Odor Stench
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 1737738
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
    Chemical Name or Material 1-Methylthio-2-propanone
    Assay ≥98%

    Description

    Applications
    1-Methylthio-2-propanone is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

    Solubility
    Soluble in alcohol and slightly soluble in water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit