Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Methylthio-2-propanone, 98+%

Click to view available options
Quantity:
5g
25g
100g
14109-72-9
C4H8OS
104.167
MFCD00015325
UKFADLGENFFWHR-UHFFFAOYSA-N
1-methylthio-2-propanone,methylthio acetone,2-thia-4-pentanone,unii-s6v0dw7bqa,1-methylsulfanyl propan-2-one,2-propanone, 1-methylthio,methylthio-2-propanone,acetonyl methyl sulfide,1-methylsulfanyl acetone,s6v0dw7bqa
6430713
1-methylsulfanylpropan-2-one
CC(=O)CSC
14109-72-9
C4H8OS
104.167
MFCD00015325
UKFADLGENFFWHR-UHFFFAOYSA-N
1-methylthio-2-propanone,methylthio acetone,2-thia-4-pentanone,unii-s6v0dw7bqa,1-methylsulfanyl propan-2-one,2-propanone, 1-methylthio,methylthio-2-propanone,acetonyl methyl sulfide,1-methylsulfanyl acetone,s6v0dw7bqa
6430713
1-methylsulfanylpropan-2-one
CC(=O)CSC
Applications
1-Methylthio-2-propanone is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in alcohol and slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Flash Point 43°C (109°F)
Boiling Point 69°C to 70°C (30mmHg)
Quantity 25g
Odor Stench
UN Number UN1224
Beilstein 1737738
Formula Weight 104.17
Percent Purity ≥98%
Chemical Name or Material 1-Methylthio-2-propanone

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.