Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-N-BOC-Aminocyclopentanecarboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass Bottle
35264-09-6
C11H19NO4
229.28
MFCD01076126
YBZCSKVLXBOFSL-UHFFFAOYSA-N
boc-cycloleucine,1-boc-amino cyclopentanecarboxylic acid,1-tert-butoxycarbonylamino cyclopentanecarboxylic acid,1-n-boc-aminocyclopentanecarboxylic acid,boc-1-aminocyclopentane-1-carboxylic acid,n-boc-cycloleucine,1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid,1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid,1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
2734645
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O
35264-09-6
C11H19NO4
229.28
MFCD01076126
YBZCSKVLXBOFSL-UHFFFAOYSA-N
boc-cycloleucine,1-boc-amino cyclopentanecarboxylic acid,1-tert-butoxycarbonylamino cyclopentanecarboxylic acid,1-n-boc-aminocyclopentanecarboxylic acid,boc-1-aminocyclopentane-1-carboxylic acid,n-boc-cycloleucine,1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid,1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid,1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
2734645
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O
Quantity 1g
Packaging Glass Bottle
Color White
Melting Point 130.0°C to 135.0°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in methanol,insoluble in toluene
Formula Weight 229.28
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-N-BOC-Aminocyclopentanecarboxylic acid
Show More Show Less