Alfa Aesar™ 1-n-Butyl-2-methyl-4-nitroimidazole, 97%

Manufacturer: Alfa Aesar™ B2489606

Quantity 5g

 View more versions of this product

Catalog No. AAB2489606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 135009-57-3
    Molecular Formula C8H13N3O2
    Formula Weight 183.21
    MDL Number MFCD00173738
    Chemical Name or Material 1-n-Butyl-2-methyl-4-nitroimidazole

    Specifications

    Formula Weight 183.21
    Molecular Formula C8H13N3O2
    MDL Number MFCD00173738
    CAS 135009-57-3
    Quantity 5g
    Chemical Name or Material 1-n-Butyl-2-methyl-4-nitroimidazole
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit