Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1-n-Butyl-2-methyl-4-nitroimidazole, 97%
Click to view available options
Quantity:
1g
5g
Specifications
Quantity | 1g |
Formula Weight | 183.21 |
Percent Purity | 97% |
Chemical Name or Material | 1-n-Butyl-2-methyl-4-nitroimidazole |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures