Alfa Aesar™ 1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene, 99+%

Manufacturer: Alfa Aesar™ H2710203

Quantity 1g

 View more versions of this product

Catalog No. AAH2710203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85583-83-1
    Molecular Formula C20H22O
    Formula Weight 278.39
    MDL Number MFCD01218016
    Synonym 1-(4-n-Butylphenyl)-2-(4-ethoxyphenyl)acetylene
    Chemical Name or Material 1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene

    Specifications

    Melting Point 54°C
    Formula Weight 278.39
    Boiling Point 168° to 170°C (0.5mmHg)
    Quantity 1g
    Molecular Formula C20H22O
    MDL Number MFCD01218016
    CAS 85583-83-1
    Synonym 1-(4-n-Butylphenyl)-2-(4-ethoxyphenyl)acetylene
    Chemical Name or Material 1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit