Alfa Aesar™ 1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene, 99+%

Manufacturer: Alfa Aesar™ H2730403

Quantity 1g

 View more versions of this product

Catalog No. AAH2730403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 35684-12-9
    Molecular Formula C19H20O
    Formula Weight 264.37
    MDL Number MFCD04038796
    Synonym 1-(4-n-Butylphenyl)-2-(4-methoxyphenyl)acetylene
    Chemical Name or Material 1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene

    Specifications

    Formula Weight 264.37
    Melting Point 40°C
    Molecular Formula C19H20O
    MDL Number MFCD04038796
    Synonym 1-(4-n-Butylphenyl)-2-(4-methoxyphenyl)acetylene
    Boiling Point 172° to 172°C (1mmHg)
    Quantity 1g
    CAS 35684-12-9
    Chemical Name or Material 1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit