Alfa Aesar™ 1-n-Butyl-4-chlorobenzene, 97%

Manufacturer: Alfa Aesar™ B2413814

Quantity 25g

 View more versions of this product

Catalog No. AAB2413814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15499-27-1
    Molecular Formula C10H13Cl
    Formula Weight 168.67
    MDL Number MFCD00051926
    Beilstein 1860506
    Synonym 4-Chloro-n-butylbenzene
    Chemical Name or Material 1-n-Butyl-4-chlorobenzene

    Specifications

    Formula Weight 168.67
    MDL Number MFCD00051926
    Boiling Point 225° to 228°C
    Quantity 25g
    Synonym 4-Chloro-n-butylbenzene
    Molecular Formula C10H13Cl
    CAS 15499-27-1
    Beilstein 1860506
    Chemical Name or Material 1-n-Butyl-4-chlorobenzene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit