Alfa Aesar™ 1-n-Heptyl-4-(4-pyridyl)pyridinium bromide, 95%

Manufacturer: Alfa Aesar™ H5559114

Quantity 25g

 View more versions of this product

Catalog No. AAH5559114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 39127-10-1
    Formula Weight 335.29
    MDL Number MFCD00134461
    Chemical Name or Material 1-n-Heptyl-4-(4-pyridyl)pyridinium bromide

    Specifications

    Formula Weight 335.29
    Melting Point 125° to 128°C
    Physical Form Powder
    MDL Number MFCD00134461
    CAS 39127-10-1
    Color White to yellow
    Quantity 25g
    Chemical Name or Material 1-n-Heptyl-4-(4-pyridyl)pyridinium bromide
    Assay 95%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit