Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-n-Propyl-4-piperidone, 98%

Click to view available options
Quantity:
25g
23133-37-1
C8H15NO
141.214
MFCD00010433
YGDZKYYCJUNORF-UHFFFAOYSA-N
1-propyl-4-piperidone,n-propyl-4-piperidone,1-propyl-piperidin-4-one,4-piperidinone, 1-propyl,1-propylpiperidin-4-on,1-propyl4-piperidone,pubchem2038,n-propyl-4-peperidone,1-n-propyl-4-piperidon,1-propyl-4-piperidinone
90006
1-propylpiperidin-4-one
CCCN1CCC(=O)CC1
23133-37-1
C8H15NO
141.214
MFCD00010433
YGDZKYYCJUNORF-UHFFFAOYSA-N
1-propyl-4-piperidone,n-propyl-4-piperidone,1-propyl-piperidin-4-one,4-piperidinone, 1-propyl,1-propylpiperidin-4-on,1-propyl4-piperidone,pubchem2038,n-propyl-4-peperidone,1-n-propyl-4-piperidon,1-propyl-4-piperidinone
90006
1-propylpiperidin-4-one
CCCN1CCC(=O)CC1
Quantity 25g
Flash Point 75°C (168°F)
Boiling Point 56°C (1mmHg)
Formula Weight 141.22
Percent Purity 98%
Chemical Name or Material 1-n-Propyl-4-piperidone

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C