Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthaldehyde, 95%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
66-77-3
C11H8O
156.18
MFCD00004003
SQAINHDHICKHLX-UHFFFAOYSA-N
1-naphthaldehyde,1-formylnaphthalene,1-naphthalenecarboxaldehyde,1-naphthylaldehyde,alpha-naphthal,alpha-naphthaldehyde,naphthaldehyde,.alpha.-naphthal,alpha-naphthylcarboxaldehyde,alpha-naphthylaldehyde
6195
CHEBI:52367
naphthalene-1-carbaldehyde
C1=CC=C2C(=C1)C=CC=C2C=O
66-77-3
C11H8O
156.18
MFCD00004003
SQAINHDHICKHLX-UHFFFAOYSA-N
1-naphthaldehyde,1-formylnaphthalene,1-naphthalenecarboxaldehyde,1-naphthylaldehyde,alpha-naphthal,alpha-naphthaldehyde,naphthaldehyde,.alpha.-naphthal,alpha-naphthylcarboxaldehyde,alpha-naphthylaldehyde
6195
CHEBI:52367
naphthalene-1-carbaldehyde
C1=CC=C2C(=C1)C=CC=C2C=O
Refractive Index 1.6500 to 1.6540
Flash Point >110°C
Melting Point 1.0°C to 2.0°C
Color Brown-Yellow to Yellow
Density 1.1500g/mL
Boiling Point 160.0°C to 161.0°C (15.0mmHg)
Specific Gravity 1.15
Quantity 2.5kg
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula C10H7CHO
Beilstein 07, 400
Solubility Information Solubility in water: <1g/L (20°C)
Formula Weight 156.18
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 1-Naphthaldehyde
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-633-4