Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthaleneacetic Acid, 97%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
86-87-3
C12H10O2
186.21
PRPINYUDVPFIRX-UHFFFAOYSA-N
2-(naphthalen-1-yl)acetic acid
OC(=O)CC1=C2C=CC=CC2=CC=C1
86-87-3
C12H10O2
186.21
PRPINYUDVPFIRX-UHFFFAOYSA-N
2-(naphthalen-1-yl)acetic acid
OC(=O)CC1=C2C=CC=CC2=CC=C1

NA107, 86-87-3, C12H10O2

Spectrum™ 1-Naphthaleneacetic Acid is an organic compound that is colorless and soluble in organic solvents. It is a synthetic plant hormone that is part of the auxin family and is found in most commercial horticultural plant rooting products and is used for rooting and propagation purposes.
Packaging Poly Pail
Quantity 1 kg
CAS Max % 100%
Percent Purity 97%