Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthaleneboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13922-41-3
C10H9BO2
171.99
HUMMCEUVDBVXTQ-UHFFFAOYSA-N
1-naphthaleneboronic acid,1-naphthylboronic acid,naphthalene-1-boronic acid,naphthalenyl-1-boronic acid,1-naphthalene boronic acid,1-naphthaleneboronicacid,1-naphthyleneboronic acid,boronic acid, naphthalenyl,1-borononaphthalene,naphthylboronic acid
254532
naphthalen-1-ylboronic acid
B(C1=CC=CC2=CC=CC=C12)(O)O
13922-41-3
C10H9BO2
171.99
HUMMCEUVDBVXTQ-UHFFFAOYSA-N
1-naphthaleneboronic acid,1-naphthylboronic acid,naphthalene-1-boronic acid,naphthalenyl-1-boronic acid,1-naphthalene boronic acid,1-naphthaleneboronicacid,1-naphthyleneboronic acid,boronic acid, naphthalenyl,1-borononaphthalene,naphthylboronic acid
254532
naphthalen-1-ylboronic acid
B(C1=CC=CC2=CC=CC=C12)(O)O
Quantity 1g
Melting Point 196.0°C to 200.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige-Pink to White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: +/- 2.5% in water.
Formula Weight 171.99
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 1-Naphthaleneboronic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning