Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthalenemethanol, 98%, ACROS Organics™

Click to view available options
Quantity:
2g
10g
Packaging:
Glass bottle
4780-79-4
C11H10O
158.2
MFCD00004044
PBLNHHSDYFYZNC-UHFFFAOYSA-N
1-naphthalenemethanol,1-naphthylmethanol,1-hydroxymethylnaphthalene,naphthalene-1-methanol,1-naphthalene methanol,1-menaphthyl alcohol,1-naphthylmethyl alcohol,naphthalen-1-yl-methanol,naphthalen-1-yl methanol,1-naphthylenemethanol
20908
CHEBI:38137
naphthalen-1-ylmethanol
C1=CC=C2C(=C1)C=CC=C2CO
4780-79-4
C11H10O
158.2
MFCD00004044
PBLNHHSDYFYZNC-UHFFFAOYSA-N
1-naphthalenemethanol,1-naphthylmethanol,1-hydroxymethylnaphthalene,naphthalene-1-methanol,1-naphthalene methanol,1-menaphthyl alcohol,1-naphthylmethyl alcohol,naphthalen-1-yl-methanol,naphthalen-1-yl methanol,1-naphthylenemethanol
20908
CHEBI:38137
naphthalen-1-ylmethanol
C1=CC=C2C(=C1)C=CC=C2CO
Boiling Point 301°C (715.0mmHg)
Color Yellow
Quantity 10g
Melting Point 59°C to 62°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C10H7CH2OH
Beilstein 06, 667
Formula Weight 158.2
Physical Form Fine Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-Naphthalenemethanol, 98%
Show More Show Less

EINECSNumber : 225-324-1

RTECSNumber : QJ8880000

TSCA : TSCA