Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthol, Purified, 96%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
90-15-3
90-15-3
Packaging Fiber Drum
Quantity 50 kg
CAS Max % 1
Percent Purity 0.96
Grade Purified

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.