Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Naphthyl butyrate, 98+%

Click to view available options
Quantity:
1g
5g
25g
3121-70-8
C14H14O2
214.264
MFCD00003923
XIVJNRXPRQKFRZ-UHFFFAOYSA-N
1-naphthyl butyrate,.alpha.-naphthyl butyrate,alpha-naphthyl butyrate,naphthalen-1-yl butyrate,butanoic acid, 1-naphthalenyl ester,butyric acid, 1-naphthyl ester,1-naphthylbutyrate,unii-9uzl9u0cik,butyric acid 1-naphthyl ester,9uzl9u0cik
76571
naphthalen-1-yl butanoate
CCCC(=O)OC1=CC=CC2=CC=CC=C21
3121-70-8
C14H14O2
214.264
MFCD00003923
XIVJNRXPRQKFRZ-UHFFFAOYSA-N
1-naphthyl butyrate,.alpha.-naphthyl butyrate,alpha-naphthyl butyrate,naphthalen-1-yl butyrate,butanoic acid, 1-naphthalenyl ester,butyric acid, 1-naphthyl ester,1-naphthylbutyrate,unii-9uzl9u0cik,butyric acid 1-naphthyl ester,9uzl9u0cik
76571
naphthalen-1-yl butanoate
CCCC(=O)OC1=CC=CC2=CC=CC=C21
Boiling Point 125°C to 130°C (1mmHg)
Flash Point 62°C (143°F)
Quantity 1g
Beilstein 2558646
Formula Weight 214.27
Percent Purity ≥98%
Chemical Name or Material 1-Naphthyl butyrate

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA