Alfa Aesar™ 1-Naphthyl butyrate, 98+%

Manufacturer: Alfa Aesar™ B2322114

Quantity 25g

 View more versions of this product

Catalog No. AAB2322114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3121-70-8
    Molecular Formula C14H14O2
    Formula Weight 214.27
    MDL Number MFCD00003923
    Beilstein 2558646
    Synonym Butyric acid 1-naphthyl ester
    Chemical Name or Material 1-Naphthyl butyrate

    Specifications

    Formula Weight 214.27
    TSCA TSCA
    Molecular Formula C14H14O2
    Boiling Point 125° to 130°C (1mmHg)
    Quantity 25g
    Flash Point 62°C (143°F)
    Synonym Butyric acid 1-naphthyl ester
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    MDL Number MFCD00003923
    CAS 3121-70-8
    Beilstein 2558646
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1-Naphthyl butyrate
    Assay ≥98%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit