Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthyl Isothiocyanate 98%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
Packaging:
Glass bottle
551-06-4
C11H7NS
185.25
MFCD00003882
JBDOSUUXMYMWQH-UHFFFAOYSA-N
1-naphthyl isothiocyanate,1-naphthylisothiocyanate,anit,kesscocide,alpha-naphthyl isothiocyanate,naphthalene, 1-isothiocyanato,naphthalene, isothiocyanato,isothiocyanic acid, 1-naphthyl ester,1-naftylisothiokyanat,isothiocyanic acid 1-naphthyl ester
11080
CHEBI:35455
1-isothiocyanatonaphthalene
C1=CC=C2C(=C1)C=CC=C2N=C=S
551-06-4
C11H7NS
185.25
MFCD00003882
JBDOSUUXMYMWQH-UHFFFAOYSA-N
1-naphthyl isothiocyanate,1-naphthylisothiocyanate,anit,kesscocide,alpha-naphthyl isothiocyanate,naphthalene, 1-isothiocyanato,naphthalene, isothiocyanato,isothiocyanic acid, 1-naphthyl ester,1-naftylisothiokyanat,isothiocyanic acid 1-naphthyl ester
11080
CHEBI:35455
1-isothiocyanatonaphthalene
C1=CC=C2C(=C1)C=CC=C2N=C=S
Color Yellow
Melting Point 55.5°C to 57.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C10H7NCS
Beilstein 12, 1244
Fieser 01,718
Merck Index 15, 6493
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ether, benzene, hot alcohol,, acetone and ccl4
Formula Weight 185.25
Physical Form Fine Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-Naphthyl isothiocyanate, 98%
Show More Show Less

GHS P Statement
Harmful in contact with skin.
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-990-8

RTECSNumber : NX9100000

TSCA : TSCA