Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthyl phosphate, calcium salt trihydrate, 97%, Thermo Scientific Chemicals

Catalog Number ac346640050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
17805-69-5
C20H16CaO8P2·3H2O
540.41
MFCD00149488
XGDPLNLBXXLFDL-UHFFFAOYSA-L
calcium naphthalen-1-yl phosphate trihydrate, calcium trihydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate calcium salt trihydrate, c10h7po4.ca.3h2o, 1-naphthyl phosphate calcium trihydrate
51346138
calcium;naphthalen-1-yl phosphate;trihydrate
C1=CC=C2C(=C1)C=CC=C2OP(=O)([O-])[O-].O.O.O.[Ca+2]
17805-69-5
C20H16CaO8P2·3H2O
540.41
MFCD00149488
XGDPLNLBXXLFDL-UHFFFAOYSA-L
calcium naphthalen-1-yl phosphate trihydrate, calcium trihydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate calcium salt trihydrate, c10h7po4.ca.3h2o, 1-naphthyl phosphate calcium trihydrate
51346138
calcium;naphthalen-1-yl phosphate;trihydrate
C1=CC=C2C(=C1)C=CC=C2OP(=O)([O-])[O-].O.O.O.[Ca+2]

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 192°C to 193°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Complexometry)
Packaging Glass bottle
Quantity 5 g
Formula Weight 540.41
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Naphthyl phosphate, calcium salt trihydrate, 97%
Show More Show Less
Hazard Category HYGROSCOPIC

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit