Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Naphthyl phosphate disodium salt hydrate, 99%

Click to view available options
Quantity:
1g
5g
25g
207569-06-0
C10H9Na2O5P
286.13
MFCD00150614
RJJPCVOIUDUWLF-UHFFFAOYSA-L
1-naphthyl phosphate disodium salt hydrate,sodium naphthalen-1-yl phosphate hydrate,disodium 1-naphthyl phosphate hydrate,disodium hydrate naphthalen-1-yl phosphate,1-naphthyl phosphate sodium salt monohydrate,c10h7o4p.2na.h2o,1-naphthyl?phosphate?disodium?salt?hydrate,1-naphthyl phosphate disodium salt monohydrate,dihydrogen naphthyl phosphate, oxamethane, sodium salt, sodium salt
24212014
disodium;naphthalen-1-yl phosphate;hydrate
C1=CC=C2C(=C1)C=CC=C2OP(=O)([O-])[O-].O.[Na+].[Na+]
207569-06-0
C10H9Na2O5P
286.13
MFCD00150614
RJJPCVOIUDUWLF-UHFFFAOYSA-L
1-naphthyl phosphate disodium salt hydrate,sodium naphthalen-1-yl phosphate hydrate,disodium 1-naphthyl phosphate hydrate,disodium hydrate naphthalen-1-yl phosphate,1-naphthyl phosphate sodium salt monohydrate,c10h7o4p.2na.h2o,1-naphthyl?phosphate?disodium?salt?hydrate,1-naphthyl phosphate disodium salt monohydrate,dihydrogen naphthyl phosphate, oxamethane, sodium salt, sodium salt
24212014
disodium;naphthalen-1-yl phosphate;hydrate
C1=CC=C2C(=C1)C=CC=C2OP(=O)([O-])[O-].O.[Na+].[Na+]
Applications
1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Melting Point >300°C
Quantity 25g
Beilstein 4895811
Formula Weight 268.12 (Anhydrous)
Percent Purity 99%
Chemical Name or Material 1-Naphthyl phosphate disodium salt hydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C