Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1-Naphthylacetamide, 98%
Click to view available options
Quantity:
25g
100g
500g
Specifications
Melting Point | 176°C to 178°C |
Quantity | 25g |
Formula Weight | 185.23 |
Percent Purity | 98% |
Chemical Name or Material | 1-Naphthylacetamide |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
EINECSNumber : 201-704-2
TSCA : Yes
Recommended Storage : Ambient temperatures