Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthylacetonitrile 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Plastic Bottle
132-75-2
C12H9N
167.211
MFCD00004041
OQRMWUNUKVUHQO-UHFFFAOYSA-N
1-naphthylacetonitrile,2-naphthalen-1-yl acetonitrile,1-naphthyl acetonitrile,1-naphthaleneacetonitrile,acetonitrile, 1-naphthyl,alpha-naphthylacetonitrile,alpha-naphthyl acetonitrile,1-naphthylmethylcyanide,.alpha.-naphthylacetonitrile,2-1-naphthyl acetonitrile
8596
2-naphthalen-1-ylacetonitrile
C1=CC=C2C(=C1)C=CC=C2CC#N
132-75-2
C12H9N
167.211
MFCD00004041
OQRMWUNUKVUHQO-UHFFFAOYSA-N
1-naphthylacetonitrile,2-naphthalen-1-yl acetonitrile,1-naphthyl acetonitrile,1-naphthaleneacetonitrile,acetonitrile, 1-naphthyl,alpha-naphthylacetonitrile,alpha-naphthyl acetonitrile,1-naphthylmethylcyanide,.alpha.-naphthylacetonitrile,2-1-naphthyl acetonitrile
8596
2-naphthalen-1-ylacetonitrile
C1=CC=C2C(=C1)C=CC=C2CC#N
Packaging Plastic Bottle
Refractive Index 1.6182 to 1.6202
Boiling Point 191.0°C to 194.0°C (18.0mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Melting Point 33.0°C to 35.0°C
Color Yellow to Brown
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C10H7CH2CN
Beilstein 09, 667
Formula Weight 167.21
Physical Form Liquid After Melting
Percent Purity 97%
Chemical Name or Material 1-Naphthylacetonitrile, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-078-1

RTECSNumber : AM1240000

TSCA : TSCA