Alfa Aesar™ 1-Naphthylamine hydrochloride, 98%

Manufacturer: Alfa Aesar™ 4408804

Quantity 2g

 View more versions of this product

Catalog No. AA4408804

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 552-46-5
    Molecular Formula C10H9N·HCl
    Formula Weight 179.65
    MDL Number MFCD00036370
    UN Number UN2811
    Merck Index 14,6398
    Synonym 1-Aminonaphthalene hydrochloride
    Chemical Name or Material 1-Naphthylamine hydrochloride

    Specifications

    Formula Weight 179.65
    Health Hazard 3 GHS P Statement
    P281-P264-P301+P312-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00036370
    Physical Form Crystalline
    Molecular Formula C10H9N·HCl
    UN Number UN2811
    Health Hazard 1 Danger
    Sensitivity Air & Light Sensitive
    Merck Index 14,6398
    Health Hazard 2 GHS H Statement
    H350-H302
    May cause cancer.
    Harmful if swallowed.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number QM3150000
    Color White to pale red
    Quantity 2g
    CAS 552-46-5
    Synonym 1-Aminonaphthalene hydrochloride
    Solubility Information Soluble in water, alcohol, ether
    Chemical Name or Material 1-Naphthylamine hydrochloride
    Assay 98%

    GHS H Statement
    H350-H302
    May cause cancer.
    Harmful if swallowed.

    GHS P Statement
    P281-P264-P301+P312-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit