Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Naphthylmethyl)triphenylphosphonium chloride, 97%, Acros Organics

Click to view available options
Quantity:
25g
23277-00-1
C29H24ClP
438.935
MOYSMPXSEXYEJV-UHFFFAOYSA-M
1-naphthylmethyl triphenylphosphonium chloride,naphthalen-1-ylmethyl triphenylphosphonium chloride,1-naphthylmethyl triphenylphosphoniumchloride,naphthalen-1-ylmethyl triphenyl phosphanium chloride,1-naphthylmethyltriphenylphosphonium chloride,acmc-209g2f,1-napthylmethyl triphenylphosphonium chloride,triphenyl 1-naphthylmethyl phosphonium chloride,1-naphthylmethyl-triphenylphosphonium chloride,1-naphthalenylmethyl triphenyl phosphonium chloride
2734113
naphthalen-1-ylmethyl(triphenyl)phosphanium;chloride
C1=CC=C(C=C1)[P+](CC2=CC=CC3=CC=CC=C32)(C4=CC=CC=C4)C5=CC=CC=C5.[Cl-]
23277-00-1
C29H24ClP
438.935
MOYSMPXSEXYEJV-UHFFFAOYSA-M
1-naphthylmethyl triphenylphosphonium chloride,naphthalen-1-ylmethyl triphenylphosphonium chloride,1-naphthylmethyl triphenylphosphoniumchloride,naphthalen-1-ylmethyl triphenyl phosphanium chloride,1-naphthylmethyltriphenylphosphonium chloride,acmc-209g2f,1-napthylmethyl triphenylphosphonium chloride,triphenyl 1-naphthylmethyl phosphonium chloride,1-naphthylmethyl-triphenylphosphonium chloride,1-naphthalenylmethyl triphenyl phosphonium chloride
2734113
naphthalen-1-ylmethyl(triphenyl)phosphanium;chloride
C1=CC=C(C=C1)[P+](CC2=CC=CC3=CC=CC=C32)(C4=CC=CC=C4)C5=CC=CC=C5.[Cl-]
for C-C Bond Formation (Olefination)
Quantity 25g
Color White
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 438.93
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material (1-Naphthylmethyl)triphenylphosphonium chloride, 97%