Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthylmethylamine 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
118-31-0
C11H11N
157.216
MFCD00004048
NVSYANRBXPURRQ-UHFFFAOYSA-N
1-naphthalenemethylamine,1-naphthalenemethanamine,1-naphthylmethylamine,1-aminomethyl naphthalene,1-naphthalenemethyl amine,c-naphthalen-1-yl-methylamine,1-naphthylmethanamine,1-aminomethylnaphthalene,naphthylmethylamine,1-naphthylmethyl amine hydrochloride
8355
naphthalen-1-ylmethanamine
C1=CC=C2C(=C1)C=CC=C2CN
118-31-0
C11H11N
157.216
MFCD00004048
NVSYANRBXPURRQ-UHFFFAOYSA-N
1-naphthalenemethylamine,1-naphthalenemethanamine,1-naphthylmethylamine,1-aminomethyl naphthalene,1-naphthalenemethyl amine,c-naphthalen-1-yl-methylamine,1-naphthylmethanamine,1-aminomethylnaphthalene,naphthylmethylamine,1-naphthylmethyl amine hydrochloride
8355
naphthalen-1-ylmethanamine
C1=CC=C2C(=C1)C=CC=C2CN
Color Yellow
Quantity 25g
Boiling Point 293°C
Formula Weight 157.22
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 1-Naphthylmethylamine