Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nitropropane 98%, ACROS Organics™

Click to view available options
Quantity:
5g
500g
Packaging:
Glass bottle
108-03-2
C3H7NO2
89.09
MFCD00007407
JSZOAYXJRCEYSX-UHFFFAOYSA-N
propane, 1-nitro,nitropropane,propane, nitro,ccris 1329,dsstox_cid_980,dsstox_rid_75902,dsstox_gsid_20980,nitro-propane,n-nitropropane,3-nitropropane
7903
CHEBI:76261
1-nitropropane
CCC[N+](=O)[O-]
108-03-2
C3H7NO2
89.09
MFCD00007407
JSZOAYXJRCEYSX-UHFFFAOYSA-N
propane, 1-nitro,nitropropane,propane, nitro,ccris 1329,dsstox_cid_980,dsstox_rid_75902,dsstox_gsid_20980,nitro-propane,n-nitropropane,3-nitropropane
7903
CHEBI:76261
1-nitropropane
CCC[N+](=O)[O-]
Density 0.9900g/mL
Refractive Index 1.4005 to 1.4025
Melting Point -108°C
Flash Point 33°C
Boiling Point 132°C
Specific Gravity 0.99
Viscosity 0.844 mPa.s (20°C)
Packaging Glass bottle
Quantity 500g
Infrared Spectrum Authentic
Water 0.1% max.
Color Undesignated
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3CH2CH2NO2
Beilstein 01, 115
Fieser 01,745
Merck Index 15, 6714
Solubility Information Solubility in water: 1.4mL/100mL (20°C). Other solubilities: miscible with many organic solvents
Formula Weight 89.09
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1-Nitropropane, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-544-9

RTECSNumber : TZ5075000

TSCA : TSCA