Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt, 90+%

Click to view available options
Quantity:
25g
100g
500g
525-05-3
C10H5NNa2O8S2
377.249
MFCD00003885
DMKMTGULLYISBH-UHFFFAOYSA-L
nitroso-r salt,nitroso r,sodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate,disodium 1-nitroso-2-naphthol-3,6-disulfonate,nitroso r salt,1-nitroso-3,6-disulfonate,2,7-naphthalenedisulfonic acid, 3-hydroxy-4-nitroso-, disodium salt,3-hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt,1-nitroso-2-naphthol-3,6-disulfonic acid disodium salt,disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulphonate
10672
disodium;3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate
C1=CC2=C(C(=C(C=C2C=C1S(=O)(=O)[O-])S(=O)(=O)[O-])O)N=O.[Na+].[Na+]
525-05-3
C10H5NNa2O8S2
377.249
MFCD00003885
DMKMTGULLYISBH-UHFFFAOYSA-L
nitroso-r salt,nitroso r,sodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate,disodium 1-nitroso-2-naphthol-3,6-disulfonate,nitroso r salt,1-nitroso-3,6-disulfonate,2,7-naphthalenedisulfonic acid, 3-hydroxy-4-nitroso-, disodium salt,3-hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt,1-nitroso-2-naphthol-3,6-disulfonic acid disodium salt,disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulphonate
10672
disodium;3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate
C1=CC2=C(C(=C(C=C2C=C1S(=O)(=O)[O-])S(=O)(=O)[O-])O)N=O.[Na+].[Na+]
Applications
1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt is the most sensitive determination of trace cobalt reagents used for determination of cobalt and potassium-sensitive reagents.

Solubility
Slightly soluble in methanol and ethanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Melting Point >300°C
Quantity 100g
Beilstein 3643930
Merck Index 14,6644
Formula Weight 377.26
Percent Purity ≥90%
Chemical Name or Material 1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QJ6480000

TSCA : TSCA