Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nitroso-2-naphthol 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
131-91-9
C10H7NO2
173.17
MFCD00003884
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
1-nitroso-2-naphthol,2-naphthalenol, 1-nitroso,nitroso-beta-naphthol,2-naphthol, 1-nitroso,1-nitroso-2-naftol,zelen moridlova 4,zelen moridlova 4 czech,alpha-nitroso-beta-naphthol,1-nitroso-2-naftol czech,unii-757i55u2qx
8580
1-nitrosonaphthalen-2-ol
C1=CC=C2C(=C1)C=CC(=C2N=O)O
131-91-9
C10H7NO2
173.17
MFCD00003884
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
1-nitroso-2-naphthol,2-naphthalenol, 1-nitroso,nitroso-beta-naphthol,2-naphthol, 1-nitroso,1-nitroso-2-naftol,zelen moridlova 4,zelen moridlova 4 czech,alpha-nitroso-beta-naphthol,1-nitroso-2-naftol czech,unii-757i55u2qx
8580
1-nitrosonaphthalen-2-ol
C1=CC=C2C(=C1)C=CC(=C2N=O)O
Melting Point 104.0°C to 108.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown
Quantity 100g
Water 0.5% max.
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ONC10H6OH
Beilstein 07, 712
Merck Index 15, 6728
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene,carbon,disulfide,caustic alkali solutions and glacial,acetic acid,slightly soluble in cold petroleum ether
Formula Weight 173.17
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 1-Nitroso-2-naphthol, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-043-0

RTECSNumber : QL4725000

TSCA : TSCA