Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nonanesulfonic acid sodium salt, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
35192-74-6
C9H19NaO3S
230.298
MFCD00025010
RUYRDULZOKULPK-UHFFFAOYSA-M
sodium nonane-1-sulfonate,sodium 1-nonanesulfonate,1-nonanesulfonic acid sodium salt,1-nonanesulfonic acid, sodium salt,sodium nonylsulfonate,sodium 1-nonanesulphonate,ipc-alks-9,1-eeie>>cea aaeni,c9h19o3s.na,potassium nonane-1-sulfonate
23664777
sodium;nonane-1-sulfonate
CCCCCCCCCS(=O)(=O)[O-].[Na+]
35192-74-6
C9H19NaO3S
230.298
MFCD00025010
RUYRDULZOKULPK-UHFFFAOYSA-M
sodium nonane-1-sulfonate,sodium 1-nonanesulfonate,1-nonanesulfonic acid sodium salt,1-nonanesulfonic acid, sodium salt,sodium nonylsulfonate,sodium 1-nonanesulphonate,ipc-alks-9,1-eeie>>cea aaeni,c9h19o3s.na,potassium nonane-1-sulfonate
23664777
sodium;nonane-1-sulfonate
CCCCCCCCCS(=O)(=O)[O-].[Na+]
Infrared Spectrum Authentic
Color White
Quantity 1g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula S(O3Na)(CH2)8CH3
Formula Weight 230.3
Physical Form Fine Powder
Percent Purity ≥96.0%
Chemical Name or Material 1-Nonanesulfonic acid sodium salt
Show More Show Less