Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nonanol, 97%, ACROS Organics™

Click to view available options
Quantity:
100mL
250mL
1L
Packaging:
Glass bottle
143-08-8
C9H20O
144.26
MFCD00002990
ZWRUINPWMLAQRD-UHFFFAOYSA-N
1-nonanol,nonanol,n-nonyl alcohol,nonyl alcohol,pelargonic alcohol,octyl carbinol,1-hydroxynonane,nonalol,n-nonanol,n-nonan-1-ol
8914
CHEBI:35986
nonan-1-ol
CCCCCCCCCO
143-08-8
C9H20O
144.26
MFCD00002990
ZWRUINPWMLAQRD-UHFFFAOYSA-N
1-nonanol,nonanol,n-nonyl alcohol,nonyl alcohol,pelargonic alcohol,octyl carbinol,1-hydroxynonane,nonalol,n-nonanol,n-nonan-1-ol
8914
CHEBI:35986
nonan-1-ol
CCCCCCCCCO
Specific Gravity 0.827
Flash Point 74°C
Melting Point -5.0°C
Infrared Spectrum Authentic
Boiling Point 213.0°C to 215.0°C
Color Colorless
Quantity 250mL
Density 0.8270g/mL
Refractive Index 1.4324 to 1.4344
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)8OH
UN Number 3082
Beilstein 01, 423
Merck Index 15, 6766
Solubility Information Solubility in water: 1g/L (20°C)
Formula Weight 144.26
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-Nonanol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-583-7