Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Nonyne, 98%

Manufacturer:  Alfa Aesar™ B2426314

Quantity 25g

 View more versions of this product

Catalog No. AAB2426314

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
1-Nonyne has been used in enantioselective synthesis of cladospolide B, C and (ent)-cladospolide D.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents, heavy metal powders.
Description & Specifications

Chemical Identifiers

3452-09-3
CH3(CH2)6C≡CH
MFCD00009567
1736573
1-Nonyne
C9H16
124.23
UN3295
n-Heptylacetylene

Specifications

33°C (91°F)
149° to 151°C
25g
C9H16
124.23
UN3295
n-Heptylacetylene
98%
Petrol-like
−50°C
3452-09-3
CH3(CH2)6C≡CH
MFCD00009567
1736573
1-Nonyne

Safety and Handling

SDS & Safety Documents

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.

GHS P Statement
P301+P330+P331-P315
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit