Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine, 98%

Click to view available options
Quantity:
5mg
25mg
91575-58-5
C26H55N2O7P
538.707
MFCD00065891
FNFHZBKBDFRYHS-RUZDIDTESA-N
mc-paf,methylcarbamyl paf,cpaf,methylcarbamyl paf c-16,methylcarbamol,methyl-carbamyl platelet-activating factor,1-o-hexadecyl-2-n-methylcarbamyl-sn-glycero-3-phosphocholine,c-paf carbamyl-paf,c-paf,mcpaf c16
124663
[(2R)-3-hexadecoxy-2-(methylcarbamoyloxy)propyl] 2-(trimethylazaniumyl)ethyl phosphate
CCCCCCCCCCCCCCCCOCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)NC
91575-58-5
C26H55N2O7P
538.707
MFCD00065891
FNFHZBKBDFRYHS-RUZDIDTESA-N
mc-paf,methylcarbamyl paf,cpaf,methylcarbamyl paf c-16,methylcarbamol,methyl-carbamyl platelet-activating factor,1-o-hexadecyl-2-n-methylcarbamyl-sn-glycero-3-phosphocholine,c-paf carbamyl-paf,c-paf,mcpaf c16
124663
[(2R)-3-hexadecoxy-2-(methylcarbamoyloxy)propyl] 2-(trimethylazaniumyl)ethyl phosphate
CCCCCCCCCCCCCCCCOCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)NC
Quantity 25mg
Color White
Formula Weight 538.71
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine

storageNote1 : -30° to -10°C